Round-Bottom Five-Neck Flasks, Vertical Necks, Ace Glass
Supplier: Ace Glass
6957-220 6957-48 6957-252 6957-20 6957-32 6957-60 6957-260 6957-222 6957-232 6957-248 6957-08 6957-208 6957-236 6957-24 6957-36 6957-52 6957-70 6957-72 6957-74 6957-76 6957-78 6957-80 6957-38 6957-238 6957-26 6957-28
89056-458EA 947.65 CAD
89056-458 89056-470 89056-872 89056-882 89056-886 89056-892 89057-138 89057-162 89057-164 89057-172 89057-178 89057-194 89057-210 89057-212 89057-222 89057-226 89083-952 89083-954 89083-956 89083-958 89083-960 89083-962 10099-022 10099-024 10767-182 10767-184
Round-Bottom Five-Neck Flasks, Vertical Necks, Ace Glass
Flasks Round Bottom Flasks
Round-bottom flasks with five necks and [ST] outer joints
The outer joints are tooled with reinforcement rings for added strength and stability. Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks.