Flat-Bottom Three-Neck Flasks, Vertical Necks, Ace Glass
Supplier: Ace Glass
6939-10 6939-15 6939-20
89056-570EA 406.75 CAD
89056-570 89056-572 89056-574
Flat-Bottom Three-Neck Flasks, Vertical Necks, Ace Glass
Flasks Round Bottom Flasks
Flat-bottom flask can sit directly on heater or stirrer without other support
The [ST] 24/40 outer joints are tooled with reinforcement rings for added strength and stability.
Ordering information: Vertical neck flasks can be ordered for angled neck capacities; contact your Avantor sales representative.