Round-Bottom Five-Neck Flasks, Threaded Port, Ace Glass
Supplier: Ace Glass
6955-08 6955-208 6955-12 6955-212 6955-215 6955-15
89056-444EA 625.12 CAD
89056-444 89056-478 89057-110 89057-122 89057-160 89057-180
Round-Bottom Five-Neck Flasks, Threaded Port, Ace Glass
Flasks Round Bottom Flasks
Round-bottom flasks with four [ST] 24/40 outer joints and #7 Ace-Thred neck
The outer joints are tooled with reinforcement rings for added strength and stability. Two of the side necks are in-line with the center neck. The remaining side necks are placed in front of the in-line necks. One neck has a [ST] outer joint and the other is internally threaded to accommodate thermometers or bleed tubes up to 7mmO.D. Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks. Supplied with #7 nylon bushing and FETFE O-ring.