Round-Bottom Four-Neck Flasks, Thermometer Joint, Ace Glass
Supplier: Ace Glass
6953-204 6953-228 6953-40 6953-12 6953-16 6953-20 6953-205 6953-216 6953-24 6953-28 6953-72 6953-212 6953-04 6953-08 6953-220 6953-208 6953-224 6953-232 6953-260 6953-32 6953-60 6953-64 6953-66
89056-830EA 569.27 CAD
89056-830 89056-852 89056-888 89057-112 89057-114 89057-116 89057-120 89057-126 89057-136 89057-140 89057-148 89057-158 89057-186 89057-188 89057-196 89057-202 89057-206 89057-208 89057-218 89057-220 89057-228 89083-944 89083-946
Round-Bottom Four-Neck Flasks, Thermometer Joint, Ace Glass
Flasks Round Bottom Flasks
Round-bottom flasks with four necks and [ST] outer joints
The outer joints are tooled with reinforcement rings for added strength and stability. Necks are vertical unless noted otherwise. Two of the side necks are in-line with the center neck. The third side neck is placed in front of the center neck and has a [ST] 10/30 outer joint that is suitable for thermometers. Heavy wall flasks feature walls that are approximately 30% heavier than standard flasks.
Ordering information: Vertical neck flasks can be ordered for angled neck capacities; contact your Avantor sales representative.
Order Now
SPECIFICATIONS
Environmentally Preferable