2-Propanol 99,8 HPLC, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol

PX1838-NP20 PX1838-1 PX1838P-1 PX1838P-6
CAPX1838-NP20EA 1351 CAD
CAPX1838-NP20 CAPX1838-1 CAPX1838P-1 CAPX1838P-6
2-Propanol 99,8 HPLC, Supelco®
2-Propanol

HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.8% min
Appearance Clear liquid, free from particulates
Carbonyl compounds (acetone or propionaldehyde) 0.002% max
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Infrared spectrum Conforms to standard
Residue after evaporation 5ppm max
Solubility in water To pass test
Titrable acid 0.0001meq/g max
Titrable base 0.0001meq/g max
UV Abs. at 210 nm 1.00AU max
UV Abs. at 220 nm 0.40AU max
UV Abs. at 230 nm 0.20AU max
UV Abs. at 245 nm 0.08AU max
UV Abs. at 260 nm 0.04AU max
UV Abs. at 275 nm 0.03AU max
UV Abs. at 300 nm 0.02AU max
UV Abs. at 350 nm 0.01AU max
UV Cut-off 210nm max
Water (H2O) 0.10% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR