Round-Bottom Four-Neck Flasks, Ace Glass Incorporated
Supplier: Ace Glass
6952-01 6952-02 6952-04 6952-08 6952-12 6952-16 6952-17 6952-18 6952-19 6952-20 6952-201 6952-202 6952-203 6952-204 6952-206 6952-208 6952-212 6952-213 6952-216 6952-217 6952-218 6952-219 6952-220 6952-224 6952-225 6952-227 6952-228 6952-229 6952-232 6952-236 6952-237 6952-24 6952-240 6952-244 6952-248 6952-25 6952-252 6952-253 6952-256 6952-257 6952-260 6952-263 6952-264 6952-266 6952-267 6952-27 6952-28 6952-29 6952-32 6952-36 6952-40 6952-44 6952-48 6952-52 6952-56 6952-60 6952-63 6952-68 6952-72 6952-73 6952-75 6952-76 6952-77 6952-78 6952-79 6952-80 6952-81 6952-82 6952-83 6952-84 6952-88 6952-89 6952-95 6952-96 6952-97 9468-11 9468-13 9468-15 9469-12 9469-14 9469-16 6952-13 6952-37 6952-03 6952-101 6952-103 6952-105 6952-107 6952-109 6952-111 6952-113 6952-115 6952-117 6952-119
89056-958EA 508.71 CAD
89056-958 89056-960 89056-962 89056-964 89056-966 89056-968 89056-970 89056-972 89056-974 89056-976 89056-978 89056-980 89056-982 89056-984 89056-986 89056-988 89056-990 89056-992 89056-994 89056-996 89056-998 89057-000 89057-002 89057-004 89057-006 89057-008 89057-010 89057-012 89057-014 89057-016 89057-018 89057-020 89057-022 89057-024 89057-026 89057-028 89057-030 89057-032 89057-034 89057-036 89057-038 89057-040 89057-042 89057-044 89057-046 89057-048 89057-050 89057-052 89057-054 89057-056 89057-058 89057-060 89057-062 89057-064 89057-066 89057-068 89057-070 89057-072 89057-074 89057-076 89057-078 89057-080 89057-082 89057-084 89057-086 89057-088 89057-090 89057-092 89057-094 89057-096 89057-098 89057-100 89057-102 89057-104 89057-106 89074-776 89074-778 89074-780 89074-782 89074-784 89074-786 89083-920 89083-922 89083-926 89083-928 89083-930 89083-932 89083-934 89083-936 89083-938 89083-940 89083-942 89083-948 89083-950
Round-Bottom Four-Neck Flasks, Ace Glass Incorporated
Flasks Round Bottom Flasks
These round-bottom flasks have four [ST] outer joints
Available with heavy or standard walls. Heavy wall flasks feature walls that are approximately 30% heavier than standard wall flasks.
The outer joints are tooled with reinforcement rings (full-sized joints only) for added strength and stability. Necks are vertical unless noted otherwise. Two of the side necks are in-line with the center neck. The third side neck is placed in front of the center neck. Fabricated from borosilicate glass.