di-Potassium hydrogen phosphate ≥98.0%, GR ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate

PX1570-7 PX1570-2 PX1570-20
CAPX1570-7EA 1945 CAD
CAPX1570-7 CAPX1570-1 CAPX1570-2 CAPX1570-20
di-Potassium hydrogen phosphate ≥98.0%, GR ACS, Supelco®
di-Potassium hydrogen phosphate

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.0% min
Chloride (Cl) 0.003% max
Color White
Form Granular powder
Heavy metals (as Pb) 5ppm max
Insoluble matter 0.01% max
Iron (Fe) 0.001% max
Loss on drying (105°C) 1.0% max
Nitrogen compounds (as N) 0.001% max
pH of a 5% solution at 25°C 8.5-9.6
Sodium (Na) 0.05% max
Sulfate (SO4) 0.005% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR