NEXTFLEX™ Cell Free DNA-Sequence Kits
Supplier: REVVITY HEALTH SCIENCES CANADA CA
NOVA-5150-01 NOVA-5150-02
CA77448-186EA 334.11 CAD
CA77448-186 CA77448-188
NEXTFLEX™ Cell Free DNA-Sequence Kits
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEXTFLEX™ cell free DNA-sequence kit 2.0 can produce libraries from 10 ng of cell free DNA for circulating tumor DNA (ctDNA) or cell free fetal DNA (cffDNA) analysis, in two hours.
  • Designed for low sample input - 10 ng of input cfDNA required
  • Accelerated workflow with minimized hands-on time
  • Flexible adapter barcode options - up to 1536 unique dual index barcodes are available
  • Compatible with Illumina® sequencing platforms
  • Protocols available for automation on the Sciclone® G3 NGSx, NGSx iQ™ and Zephyr® G3 NGS workstations
The NEXTFLEX™ cell free DNA-sequence kit 2.0 delivers high coverage and reduced bias for Illumina® sequencing applications. The NEXTFLEX™ cell free DNA-sequence kit 2.0 can be used to prepare single, paired-end and multiplexed DNA libraries for sequencing using Illumina® platforms. The availability of up to 1536 unique dual index barcodes facilitates high-throughput applications without compromising data integrity.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.