I-CHEM™ Cylinder Round Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
013-0500BP 013-1000BP 013-0250BP
IR-013-0500BPCS 285.25 CAD
IR-013-0500BP IR013-1000BP IR013-0250BP
I-CHEM™ Cylinder Round Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Water Sampling Bottles
Thermo Scientific HDPE Cylinder Rounds are general-purpose containers, supplied bulk and unassembled.
Unprocessed, ready for your own cleaning procedure and assembly with caps purchased separately. These are the same high quality container components as the assembled HDPE cylinder rounds, but sold in bulk, without a cap.
Ordering information: Bulk packed. Closures sold separately, see IR067-2410WBP series.