Sulfuric acid 95-98% ACS, FCC, VWR Chemicals BDH®
Supplier: VWR International
|
Danger
|
CABDH3090-185L
CABDH3068P
CABDH3070PC
CABDH30722.5LG
CABDH30743.8LP
CABDH3132-2.2P
CABDH3090-185LEA
3472.12
CAD
CABDH3090-185L
CABDH3068P
CABDH3070PC
CABDH30722.5LG
CABDH30743.8LP
CABDH3132-2.2P
Sulfuric acid 95-98% ACS, FCC, VWR Chemicals BDH®
Sulfuric acid
Free from particulates.
Meets reagent specifications for testing in FCC monographs.
Formula:
H₂SO₄ MW: 98.08 Boiling Pt: 330 Melting Pt: 10.38 Density: 1.84 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8 II |
Specification Test Results
Ammonium (NH4) | 2 ppm max. |
Appearance | Clear, colorless solution |
Arsenic (As) | 0.01 ppm max. |
Assay (H2SO4) | 95.0 - 98.0% |
Chloride (as Cl) | 0.2 ppm max. |
Color (APHA) | 10 max. |
Free from suspended matter | To pass test |
Heavy metals (as Pb) | 1 ppm max. |
Identification: sulfate per FCC | To pass test |
Iron (Fe) | 0.2 ppm max. |
Mercury (Hg) | 5 ppb max. |
Nitrate (NO3) | 0.5 ppm max. |
Residue after ignition | 5 ppm max. |
Selenium (Se) | 0.002% max. |
Substances reducing permanganate (ACS Method) | To pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...