VWR® Traceable® Jumbo-Display Dial Thermometers
Supplier: VWR International
90028-45 93677-18
61161-310EA 142.3 CAD
61161-310 77776-720
VWR® Traceable® Jumbo-Display Dial Thermometers
Thermometers Digital Thermometers
Thermometers fit cuvettes, test tubes, flasks, and beakers and will read temperatures in samples, reagents, water baths, foods, gases, and semi-solids.
  • Traceable®
  • Wide temperature range
  • Easy-to-read LCD
  • Read °C and °F
Readings update every second. Stainless steel probe is resistant to most laboratory chemicals. Dial has 5.4 cm (21/8") diameter and features a large, bright 1 cm (3/8") high LCD.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with replaceable 1.5V silver-oxide battery.