VWR® TraceClean® Wide Mouth Jars, Glass
Supplier: VWR International
89093-942 89093-944 89093-950 89093-958 89093-960 89093-992 89093-994 89093-996 89093-998 89094-000 89094-002 89094-004 89094-006 89094-008 89094-010 89094-012 89094-014 89093-952
89093-942CS 255.15 CAD
89093-942 89093-944 89093-950 89093-958 89093-960 89093-992 89093-994 89093-996 89093-998 89094-000 89094-002 89094-004 89094-006 89094-008 89094-010 89094-012 89094-014 89093-952
VWR® TraceClean® Wide Mouth Jars, Glass
Jars General Use Jars
Type III (soda-lime) jars are for collection of solid matrices or liquids dispensed from bailers or peristaltic tubing.
  • Soda-lime glass fabrication
  • Wide-mouthed design
  • Available with solid-top or open-top caps
  • Individually numbered for traceability
  • Jars are available with solid-top or open-top caps
Processed (PC) and Quality-Assured® (QA) jars follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA jars include a Certificate of Analysis in every case.