VWR® TraceClean® Cubitainer Containers, LDPE
Supplier: VWR
VWR® TraceClean®
QLPC15900216 QLPC15900310 QLPC15900126 QLPC15900124 QLPC15900218
89094-142CS 470.06 CAD
89094-142 89094-144 89094-140 89094-136 89094-138
VWR® TraceClean® Cubitainer Containers, LDPE
Bottles Water Sampling Bottles
Containers are for collecting groundwater and influent, effluent, and bio-assay waters.
  • Low-density polyethylene construction
  • Completely traceable
  • Custody-sealed to ensure content integrity
  • Supplied with 38-400 polyfoam-lined caps
Processed (PC) and Quality-Assured® (QA) containers follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'.
QA containers include a Certificate of Analysis in every case, as well as bar codes with lot number and individual container number for complete traceability. Not Processed (NPC) containers do not receive the specialty cleaning process. All cases are custody-sealed to ensure content integrity.