VWR® TraceClean® Cylinder Bottles, HDPE
Supplier: VWR International
89094-072 89094-074 89094-076 89094-080 89094-082 89094-084 89094-088 89094-090 89094-092
89094-072CS 151.94 CAD
89094-072 89094-074 89094-076 89094-080 89094-082 89094-084 89094-088 89094-090 89094-092
VWR® TraceClean® Cylinder Bottles, HDPE
Bottles Water Sampling Bottles
Plastic bottles are for use in metals analyses and multiple inorganic analytes.
  • HDPE construction
  • Suited to metals analyses
  • Supplied with polyfoam-lined caps
Processed (PC) and Quality-Assured® (QA) bottles follow EPA cleaning procedures listed in the OSWER Directive 9240.0-05A 'Specifications and Guidance for Contaminant-Free Sample Containers'. QA bottles include a Certificate of Analysis in every case, as well as bar codes with lot number and individual container number for complete traceability.
Not Processed (NPC) bottles do not receive theMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable