Memphis Glove® Survivor™ Cut-Resistant Gloves, MCR Safety
Supplier: MCR Safety
9345LD 9345MD 9345XSD
80081-616EA 62.43 CAD
80081-616 80081-618 80081-624
Memphis Glove® Survivor™ Cut-Resistant Gloves, MCR Safety
Gloves
Ideal for use in food processing applications, these gloves are constructed of FDA-approved cut-resistant Spectra® and synthetic blend fiber materials.
  • Ambidextrous
  • Reversible
  • Knit wrist
Gloves can be laundered without shrinkage.
Ordering information: Gloves are sold as singles.