AlphaTec® 02-100 Chemical resistant gloves, Ansell
Supplier: Ansell Canada
0210011060 0210011070 0210011080 0210011090 0210011110
CA89041-814CS 1177.25 CAD
CA89041-814 CA89041-816 CA89041-818 CA89041-820 CA89041-824
AlphaTec® 02-100 Chemical resistant gloves, Ansell
Style 02-100 flat-film gloves are constructed of a five-layer composite of polyethylene, nylon, and metallocene.
  • Can be worn beneath heavier gloves
  • Useful for handling in many different industries
  • Greater dexterity with winged thumbs
  • Hand-specific
The unique polymer and seam-fusion technology provide a secure seal against a wide range of chemicals, as well as a flexible, comfortable fit.
Uses include aircraft assembly; chemical processing, oil refining and petrochemicals; handling acids, alkalies, salts, ketones and alcohols; handling strong organic solvents; chemical plants and transportation; graphic arts and printing cleanup; handling specialty chemicals; and HazMat work. Can be worn as a liner under heavier gloves. Each glove is individually air-pressure tested and are made of HPPE.
Packaging: Gloves are packaged 12 pairs in a bag; 72 pairs in a carton.