Silicon dioxide ≥99.995% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
88777-14
88777-22
CAAA88777-14EA
176.96
CAD
CAAA88777-14
CAAA88777-22
Silicon dioxide ≥99.995% (metals basis)
Silicium dioxide
Powder, 40 mesh. Purity based on metals contaminates only.
Purity: 99.995%
CAS No.: 7631-86-9
Molecular Formula: SiO2
Formula Weight: 60.09
Physical Form: -40 Mesh Powder
Density: 2.2
Appearance: White
Odor: Odorless
Melting Point: 1710°
Boiling Point: 2230°
Merck Reference: 14,8493
MDL No.: MFCD00011232
Formula:
SiO₂ MW: 60.08 g/mol Boiling Pt: 2230 Melting Pt: 1719 Density: 2.2 Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 7631-86-9 EINECS: 231-545-4 Merck Index: 14,08493 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...