Selenious acid 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Selenous acid

12574-06 12574-30
CAAA12574-06AEA 31.28 CAD
CAAA12574-06A CAAA12574-30A CAAA12574-18A
Selenious acid 97%
Selenious acid

As an oxidizing agent, as a reagent for alkaloids


Purity: 97% CAS No.: 7783-00-8 Molecular Formula: H2SeO3 Formula Weight: 128.97 Physical Form: -4 Mesh Sensitivity: Hygroscopic Solubility: Freely soluble in water, alcohol. Insoluble in ammonia Merck Reference: 14,8430 UN No.: 3290 MDL No.: MFCD00011331


As an oxidizing agent, as a reagent for alkaloids

Formula: H₂O₃Se
MW: 128.97
Boiling Pt: 315
Melting Pt: 70 (dec.)
Density: 1.998 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011331
CAS Number: 7783-00-8
EINECS: 231-974-7
UN: 3290
ADR: 6.1 II
Merck Index: 14,08430

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR