Polyethylene glycol 400, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms:
PEG 400
PX1286B-2
PX1286B-20
CAPX1286B-2EA
322.94
CAD
CAPX1286B-2
CAPX1286B-20
Polyethylene glycol 400, Sigma-Aldrich®
Polyethylene glycol 400
For Synthesis.
Formula:
H(OCH₂CH₂)nOH MW: 400 g/mol Boiling Pt: ∼250 °C (1013 hPa) Melting Pt: 4…8 °C Density: 1.13 g/cm³ (20 °C) Flash Pt: 245 Storage Temperature: Ambient |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 EINECS: 500-038-2 |
Specification Test Results
Clarity | To pass test |
Color (APHA) | 18 max |
Form | Liquid |
Infrared spectrum | Conforms to standard |
pH of a 5% solution at 25°C | 4.5-7.5 |
Sulfated ash | 0.05% max |
Suspended matter | To pass test |
Water solubility | To pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...