Discandium trioxide ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

Synonyms: Scandium oxide, Scandium (III) oxide

11216-03 11216-06 11216-14
CAAA11216-03EA 97.8 CAD
CAAA11216-03 CAAA11216-06 CAAA11216-14
Discandium trioxide ≥99.9% (REO, rare earth oxide basis), REacton®
Discandium trioxide

Powder. Purity based on rare earth contaminates.


Purity: 99.9% CAS No.: 12060-08-1 Molecular Formula: Sc2O3 Formula Weight: 137.91 Physical Form: Powder Refractive Index: 1.964 Odor: Odorless Melting Point: 2485° Merck Reference: 14,8392 MDL No.: MFCD00011223

Formula: O₃Sc₂
MW: 137.91 g/mol
Melting Pt: 2485 °C
Density: 3.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011223
CAS Number: 12060-08-1
EINECS: 235-042-0
Merck Index: 14,8392

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR