Sodium chloride ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
Warning
|
10862-14
10862-22
10862-36
CAAA10862-14EA
116.45
CAD
CAAA10862-14
CAAA10862-22
CAAA10862-36
Sodium chloride ≥99.998% (metals basis), Puratronic®
Sodium chloride
Powder, crystalline. Hygroscopic. Purity based on metal contaminates only. Density reading taken at 25°C.
Purity: 99.999%
CAS No.: 7647-14-5
Molecular Formula: ClNa
Formula Weight: 58.44
Physical Form: Crystalline
Density: 2.165
Refractive Index: 1.5442
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
pH: 6-9
Melting Point: 801°
Boiling Point: 1413°
Merck Reference: 14,8599
MDL No.: MFCD00003477
Formula:
NaCl Boiling Pt: 1413 °C Melting Pt: 801 °C Density: 2.165 Storage Temperature: Ambient |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 EINECS: 231-598-3 Merck Index: 14,8599 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...