Sodium carbonate, anhydrous ≥99.997% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
10861-06
10861-14
10861-22
10861-36
CAAA10861-06EA
98.28
CAD
CAAA10861-06
CAAA10861-14
CAAA10861-22
CAAA10861-36
Sodium carbonate, anhydrous ≥99.997% (metals basis), Puratronic®
Sodium carbonate
Powder. Hygroscopic. Purity based on metal contaminates only.
Purity: 99.997%
CAS No.: 497-19-8
Molecular Formula: Na2CO3
Formula Weight: 105.99
Physical Form: Powder
Density: 2.53
Refractive Index: 1.535
Sensitivity: Hygroscopic
Odor: Odorless
pH: 11,5
Beilstein Reference: 4154566
Merck Reference: 14,8596
MDL No.: MFCD00003494
Formula:
Na₂CO₃ Boiling Pt: 1600 °C (1013 hPa) Melting Pt: 854 °C Density: 2.53 Storage Temperature: Ambient |
MDL Number:
MFCD00003494 CAS Number: 497-19-8 EINECS: 207-838-8 REACH: 01-2119485498-19 Merck Index: 14,8596 |
|
Specification Test Results
Assay (by Metals Basis) | 99.997% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...