Selenium ≥99.999% (metals basis), amorphous shot, Puratronic® 2-6 mm (0.08-0.2 in)

Supplier: Thermo Scientific Chemicals
Danger

10603-18 10603-30 10603-A1
CAAA10603-18EA 143.94 CAD
CAAA10603-18 CAAA10603-30 CAAA10603-A1
Selenium ≥99.999% (metals basis), amorphous shot, Puratronic® 2-6 mm (0.08-0.2 in)
Selenium

MDL: MFCD00134090


Purity: 99.999% CAS No.: 7782-49-2 Odor: Odorless MDL No.: MFCD00134090

Formula: Se
Boiling Pt: ∼685 °C (1013 hPa)
Melting Pt: 217 °C
Density: 4.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134090
CAS Number: 7782-49-2
EINECS: 231-957-4
UN: 3283
ADR: 6.1,III
Merck Index: 12,08572

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR