Sulfuric acid 93-98%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®

Supplier: BDH
Danger

10301-VWNQ03 10301-VWNQ09
87003-271EA 277.26 CAD
87003-271 87003-273
Sulfuric acid 93-98%, ARISTAR® PLUS for trace metal analysis, VWR Chemicals BDH®
Sulfuric acid

High purity acid for quantitative trace metal analysis at the parts-per-billion (ppb) level.

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Assay 93 - 98 %
Color max. 10 APHA
Aluminum (Al) max. 1 ppb
Antimony (Sb) max. 1 ppb
Arsenic (As) max. 0.5 ppb
Barium (Ba) max. 0.1 ppb
Beryllium (Be) max. 0.1 ppb
Bismuth (Bi) max. 0.1 ppb
Cadmium (Cd) max. 0.5 ppb
Calcium (Ca) max. 1 ppb
Cerium (Ce) max. 0.1 ppb
Cesium (Cs) max. 0.1 ppb
Chromium (Cr) max. 0.5 ppb
Cobalt (Co) max. 0.5 ppb
Copper (Cu) max. 0.5 ppb
Dysprosium (Dy) max. 0.1 ppb
Erbium (Er) max. 0.1 ppb
Europium (Eu) max. 0.1 ppb
Gadolinium (Gd) max. 0.1 ppb
Gallium (Ga) max. 0.1 ppb
Germanium (Ge) max. 1 ppb
Gold (Au) max. 0.5 ppb
Hafnium (Hf) max. 0.1 ppb
Holmium (Ho) max. 0.1 ppb
Indium (In) max. 0.1 ppb
Iron (Fe) max. 1 ppb
Lanthanum (La) max. 0.1 ppb
Lead (Pb) max. 0.1 ppb
Lithium (Li) max. 0.5 ppb
Lutetium (Lu) max. 0.1 ppb
Magnesium (Mg) max. 1 ppb
Manganese (Mn) max. 0.5 ppb
Mercury (Hg) max. 0.1 ppb
Molybdenum (Mo) max. 0.5 ppb
Neodymium (Nd) max. 0.1 ppb
Nickel (Ni) max. 0.5 ppb
Niobium (Nb) max. 0.1 ppb
Palladium (Pd) Information Only
Platinum (Pt) Information Only
Potassium (K) max. 1 ppb
Praseodymium (Pr) max. 0.1 ppb
Rhodium (Rh) max. 0.5 ppb
Rubidium (Rb) max. 0.5 ppb
Samarium (Sm) max. 0.1 ppb
Scandium (Sc) max. 0.1 ppb
Selenium (Se) max. 10 ppb
Silver (Ag) max. 1 ppb
Sodium (Na) max. 1 ppb
Strontium (Sr) max. 0.5 ppb
Tantalum (Ta) Information Only
Tellurium (Te) max. 0.1 ppb
Terbium (Tb) max. 0.1 ppb
Thallium (Tl) max. 0.1 ppb
Thorium (Th) max. 0.1 ppb
Thulium (Tm) max. 0.1 ppb
Tin (Sn) max. 1 ppb
Titanium (Ti) max. 1 ppb
Tungsten (W) max. 0.5 ppb
Uranium (U) max. 0.1 ppb
Vanadium (V) max. 0.5 ppb
Ytterbium (Yb) max. 0.1 ppb
Yttrium (Y) max. 0.1 ppb
Zinc (Zn) max. 1 ppb
Zirconium (Zr) max. 0.5 ppb
Chloride (Cl-) max. 0.7 ppm
Nitrate (NO3-) max. 0.2 ppm
Substances Reducing Permanganate (KMnO4) max. 20 ppm
Total Phosphorus (P) max. 0.05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR