Orthophosphoric acid ≥85-88% (w/w), Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

PX1000-3 PX1000-6
CAPX1000-3CS 1796 CAD
CAPX1000-3 CAPX1000-6
Orthophosphoric acid ≥85-88% (w/w), Supelco®
Orthophosphoric acid
Formula: H₃PO₄
Melting Pt: 28 °C
Density: 1.71…1.87 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011340
CAS Number: 7664-38-2
EINECS: 231-633-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Alkali phosphates To pass test
Arsenic (As) 0.001% max
Assay 85-88%
Color (APHA) 40 max
Description To pass test
Form Clear liquid
Heavy metals (as Pb) 0.002% max
Identification To pass test
Nitrate To pass test
Phosphorous or hypophosphorous acid To pass test
Solubility To pass test
Sulfate To pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR