Orthophosphoric acid ≥85% (w/w), GR ACS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ300
PX0995-78
PX0995-3
PX0995-4000
PX0995-6
PX0995P-1
PX0995P-5
CAPX0995-78EA
2131
CAD
CAPX0995-78
CAPX0995-3
CAPX0995-4000
CAPX0995-6
CAPX0995P-1
CAPX0995P-5
Orthophosphoric acid ≥85% (w/w), GR ACS, Supelco®
Orthophosphoric acid
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Formula:
H₃PO₄ Melting Pt: 28 °C Density: 1.71…1.87 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011340 CAS Number: 7664-38-2 EINECS: 231-633-2 UN: 1805 ADR: 8,III |
Specification Test Results
Antimony (Sb) | 0.002% max |
Arsenic (As) | 1ppm max |
Assay | 85.0% min |
Calcium (Ca) | 0.002% max |
Chloride (Cl) | 3ppm max |
Color (APHA) | 10 max |
Form | Clear liquid |
Heavy metals (as Pb) | 0.001% max |
Insoluble matter | 0.001% max |
Iron (Fe) | 0.003% max |
Magnesium (Mg) | 0.002% max |
Manganese (Mn) | 0.5ppm max |
Nitrate (NO3) | 5ppm max |
Potassium (K) | 0.005% max |
Reducing substances | To pass test |
Sodium (N/A) | 0.025% max |
Sulfate (SO4) | 0.003% max |
Volatile acids (as acetic acid) | 0.001% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...