Rubidium ≥99.75% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger


Breakseal ampoule

10315-03 10315-06 10315-14 10315-22
CAAA10315-03EA 206.98 CAD
CAAA10315-03 CAAA10315-06 CAAA10315-14 CAAA10315-22
Rubidium ≥99.75% (metals basis)
Rubidium

Purity based on metal analysis only.


Purity: 99.75% CAS No.: 7440-17-7 Physical Form: Vacuum sealed in breakseal ampoules Appearance: Silver-white UN No.: 1423 MDL No.: MFCD00134055

Formula: Rb
MW: 85.47 g/mol
Boiling Pt: 700 °C (1013 hPa)
Melting Pt: 39 °C
Density: 0.83 g/cm³ (1399.85 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134055
CAS Number: 7440-17-7
EINECS: 231-126-6
UN: 1423
ADR: 4.3,I
Merck Index: 12,08439

Order Now

Specification Test Results

Assay (by Metals Basis) 99.75%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR