2,7-Octadienol (cis- and trans- mixture) ≥95.0%
Supplier: TCI America
O0243-25ML
O0243-500ML
TCO0243-025MLEA
62.68
CAD
TCO0243-025ML
TCO0243-500ML
2,7-Octadienol (cis- and trans- mixture) ≥95.0%
2,7-Octadienol (cis- and trans- mixture)
CAS Number: 23578-51-0
MDL Number: MFCD00191472
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 83
Specific Gravity (20/20): 0.87
Formula:
C₈H₁₄O Density: 0.87 (20 °C) Flash Pt: 83 °C |
MDL Number:
MFCD00191472 CAS Number: 23578-51-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...