Tetrahydro-4H-thiopyran-4-one ≥98.0%

Supplier: TCI America

Synonyms: Tetrahydrothiopyran-4-one

O0112-5G O0112-25G
TCO0112-005GEA 109.02 CAD
TCO0112-005G TCO0112-025G
Tetrahydro-4H-thiopyran-4-one ≥98.0%
Tetrahydro-4H-thiopyran-4-one

CAS Number: 1072-72-6
MDL Number: MFCD00006660
Molecular Formula: C5H8OS
Molecular Weight: 116.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 63


Other Product Names / Synonyms: Tetrahydrothiopyran-4-one

Formula: C₅H₈OS
MW: 116.18 g/mol
Melting Pt: 63 °C
MDL Number: MFCD00006660
CAS Number: 1072-72-6

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR