1-Octanesulfonic acid sodium salt ≥98.0%

Supplier: TCI America
Warning

Synonyms: Sodium octane-1-sulfonate, Sodium octane-1-sulphonate

I0346-5G I0346-25G I0346-100G
TCI0346-005GEA 45.82 CAD
TCI0346-005G TCI0346-025G TCI0346-100G
1-Octanesulfonic acid sodium salt ≥98.0%
1-Octanesulphonic acid sodium salt

[Reagent for Ion-Pair Chromatography]
CAS Number: 5324-84-5
MDL Number: MFCD00007544
Molecular Formula: C8H18O3S
Molecular Weight: 216.27
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples
Other Product Names / Synonyms: IPC-ALKS-8, 1-Octanesulfonic Acid Sodium Salt

Formula: C₈H₁₇NaO₃S
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10fcb68d113d483d32a75081b0621e88924eb676&vid=c38540471dab9c70cb65231474fd05e8b93b0d38&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR