Octyl phenol ethoxylate, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

BAKRX198-05 BAKRX198-07 BAKRX198-09
CAJTX198-5EA 173.41 CAD
CAJTX198-5 CAJTX198-7 CAJTX198-9
Octyl phenol ethoxylate, BAKER ANALYZED®, J.T.Baker®
Octyl phenol ethoxylate

Caution: Keep container tightly closed. Minimum recommended storage temperature is 23 °C.
Note: A precipitate may form on prolonged storage. Heat to 40 to 50° C with agitation to render homogeneous solution.

MDL Number: MFCD00128254
CAS Number: 9036-19-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Color (APHA) ≤125
Specific Gravity at 25°/25°C 1.055 - 1.070
Keep container tightly closed. Minimum recommended storage temperature
is 73° F (23° C). Note: A precipitate may form on prolonged storage.
Heat to 40° – 50° C with agitation to render homogeneous solution.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR