1-Octanol ≥98%, Supelco®
Supplier: MilliporeSigma
|
Warning
|
OX0130-1
OX0130-3
CAOX0130-1CS
2684
CAD
CAOX0130-1
CAOX0130-3
1-Octanol ≥98%, Supelco®
1-Octanol
Formula:
H₃C(CH₂)₆CH₂OH Boiling Pt: 195 °C (1013 hPa) Melting Pt: –16 °C Density: 0.8246 g/cm³ (20 °C) Flash Pt: 81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002988 CAS Number: 111-87-5 EINECS: 203-917-6 Merck Index: 13,06784 |
|
Specification Test Results
Assay (GC) | 98% min |
Color (APHA) | 10 max |
Form | Clear liquid |
Infrared spectrum | Conforms to standard |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...