Iodine 0.05M pyridine Oxidizing reagent, Biosynthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

BI0424-NP20 BI0424-1005 BI0424-2505 BI0424-4000 BI0424-P4000
CABI0424-NP20EA 9955 CAD
CABI0424-NP20 CABI0424-1005 CABI0424-2505 CABI0424-4000 CABI0424-P4000
Iodine 0.05M pyridine Oxidizing reagent, Biosynthesis, Sigma-Aldrich®
Iodine

Oxidizing reagents are involved in the formulations for oligonucleotide synthesis


These are suitable for all kind of biosynthesis processes such as DNA, RNA and peptide synthesis.

Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 1282
ADR: 3 II
Merck Index: 13,05036

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR