Oxidation reagent 0.05 M I₂ Pyridine/Water_90/10_vv, BioSyn™ Biochemistry, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Warning

BR664-4 BR664-4X2.5GL
CABJBR664-4CS 4525.12 CAD
CABJBR664-4 CABJBR664425GL
Oxidation reagent 0.05 M I₂ Pyridine/Water_90/10_vv, BioSyn™ Biochemistry, Burdick & Jackson™
Iodine

Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • Ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3 (main component)
Melting Pt: 114 (main component)
Density: 0.983 g/cm3 (20 °C) (main component)
Flash Pt: 12.4 %(V) (main component)
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
UN: 1993
ADR: 3 III
Merck Index: 13,05036

Order Now

Specification Test Results

Iodine 0.04 - 0.06M
Pyridine (v/v) 89.0 - 91.0%
Water (v/v) 9.0 - 11.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR