Dicobalt octacarbonyl stabilized

Supplier: Thermo Scientific Chemicals
Danger


Stabilization: Stabilised with hexane 1-5%

13060-14 13060-22 13060-88
CAAA13060-14EA 431.34 CAD
CAAA13060-14 CAAA13060-22 CAAA13060-88
Dicobalt octacarbonyl stabilized
Dicobalt octacarbonyl

Crystals. Contains 1–5% hexane as a stabilizer.


CAS No.: 10210-68-1 Molecular Formula: C8Co2O8 Formula Weight: 341.95 Flash Point: -13°(-9°F) Physical Form: Crystalline Density: 1.73 Sensitivity: Air Sensitive Appearance: Red-orange Melting Point: ca 51° dec. Solubility: Insoluble in water. Soluble in alcohol, ether and carbon disulfide UN No.: 3124 MDL No.: MFCD00016024


Applications: As a catalyst in oxo process, and in high-purity cobalt salts

Formula: C₈Co₂O₈
Melting Pt: ∼ 51 °C (dec.)
Density: 1.73
Flash Pt: –13 °C (9 °F)
Storage Temperature: Ambient
MDL Number: MFCD00016024
CAS Number: 10210-68-1
EINECS: 233-514-0
UN: 3124
ADR: 6.1,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR