3-Nitrophthalonitrile ≥98.0%

Supplier: TCI America

N0566-5G N0566-25G
TCN0566-005GEA 102.7 CAD
TCN0566-005G TCN0566-025G
3-Nitrophthalonitrile ≥98.0%
3-Nitrophthalonitrile

CAS Number: 51762-67-5
MDL Number: MFCD00191558
Molecular Formula: C8H3N3O2
Molecular Weight: 173.13
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 163


Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: 1,2-Dicyano-3-nitrobenzene

Formula: C₈H₃N₃O₂
Melting Pt: 163 °C
MDL Number: MFCD00191558
CAS Number: 51762-67-5
UN: 3276
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a45af161efb3840a7faed97acde95f0b88e0869f&vid=a45af161efb3840a7faed97acde95f0b88e0869f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR