New methylene Blue N for reticulocyte count

Supplier: Ricca Chemical

R5285000-500A R5285000-120A
RC528516EA 306.52 CAD
RC528516 RC52854
New methylene Blue N for reticulocyte count
New methylene Blue N

For reticulocytes.

Formula: C₁₈H₂₂ClN₃S
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00135878
CAS Number: 1934-16-3

Order Now

Specification Test Results

Absorbance (traceable to NIST SRM 930) Conforms to control
Appearance Deep, purple odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR