Nitric acid 10%, StandARd®, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Nitric acid solution

Management of Change (MOC) category = N

H262-43 H262-01 MACRH262-01 MACRH262-43
CAMKH26243EA 295.59 CAD
MKH26201 MKH26243 CAMKH26243 CAMKH26201
Nitric acid 10%, StandARd®, Macron Fine Chemicals™
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
MDL Number: MFCD00011349
CAS Number: 7697-37-2
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (HNO₃)(g/100 mL) 9.5 - 10.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR