Nitric acid 69,0 - 70,0% NF, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Nitric acid solution

Management of Change (MOC) category = R

BAKR9607-33 9607-04
CAJT9607-33CS 7169.9 CAD
CAJT9607-33 JT9607-4
Nitric acid 69,0 - 70,0% NF, J.T.Baker®
Nitric acid

Corrosive.

Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (as HNO₃) 69.0 - 70.0 %
NF - Clarity and Color Passes Test
NF - Identification Passes Test
NF - Chloride (Cl) ≤0.5 ppm
NF - Sulfate (SO₄) ≤1 ppm
NF - Iron (Fe) ≤0.2 ppm
NF - Residue on Ignition ≤5 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR