Nitric acid 69.0 - 70.0%, CMOS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Nitric acid solution
Management of Change (MOC) category = R
9606-03
9606-03
JT9606-3CS
520.44
CAD
JT9606-3
CAJT9606-3
Nitric acid 69.0 - 70.0%, CMOS, J.T.Baker®
Nitric acid
Formula:
HNO₃ MW: 63.01 g/mol Boiling Pt: 120.5 °C (1013 hPa) Melting Pt: –42 °C Density: 1.4 g/cm³ (20 °C) |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 UN: 2031 ADR: 8,II Merck Index: 13,06608 |
Specification Test Results
Assay (HNO₃} | 69.0 - 70.0 % |
Appearance Passes Test | Passes Test |
Color (APHA) | ≤10 |
Residue after Ignition | ≤2 ppm |
Chloride (Cl) | ≤0.08 ppm |
Phosphate (PO₄) | ≤0.1 ppm |
Sulfate (SO₄) | ≤0.2 ppm |
Trace Impurities - Aluminum (Al) | ≤40.0 ppb |
Arsenic and Antimony (as As) | ≤5 ppb |
Trace Impurities - Barium (Ba) | ≤10.0 ppb |
Trace Impurities - Beryllium (Be) | ≤10.0 ppb |
Trace Impurities - Bismuth (Bi) | ≤20.0 ppb |
Trace Impurities - Boron (B) | ≤10.0 ppb |
Trace Impurities - Cadmium (Cd) | ≤50.0 ppb |
Trace Impurities - Calcium (Ca) | ≤50.0 ppb |
Trace Impurities - Chromium (Cr) | ≤30.0 ppb |
Trace Impurities - Cobalt (Co) | ≤10.0 ppb |
Trace Impurities - Copper (Cu) | ≤10.0 ppb |
Trace Impurities - Gallium (Ga) | ≤10.0 ppb |
Trace Impurities - Germanium (Ge) | ≤20.0 ppb |
Trace Impurities - Gold (Au) | ≤20.0 ppb |
Heavy Metals (as Pb) | ≤100 ppb |
Trace Impurities - Iron (Fe) | ≤40.0 ppb |
Trace Impurities - Lead (Pb) | ≤20.0 ppb |
Trace Impurities - Lithium (Li) | ≤10.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤20.0 ppb |
Trace Impurities - Manganese (Mn) | ≤10.0 ppb |
Trace Impurities - Nickel (Ni) | |
Trace Impurities - Niobium (Nb) | |
Trace Impurities - Potassium (K) | ≤50.0 ppb |
Trace Impurities - Silicon (Si) | ≤50.0 ppb |
Trace Impurities - Silver (Ag) | ≤20.0 ppb |
Trace Impurities - Sodium (Na) | ≤150.0 ppb |
Trace Impurities - Strontium (Sr) | ≤30.0 ppb |
Trace Impurities - Tantalum (Ta) | ≤10.0 ppb |
Trace Impurities - Thallium (Tl) | ≤10.0 ppb |
Trace Impurities - Tin (Sn) | ≤20.0 ppb |
Trace Impurities - Titanium (Ti) | ≤10.0 ppb |
Trace Impurities - Vanadium (V) | ≤10.0 ppb |
Trace Impurities - Zinc (Zn) | ≤20.0 ppb |
Trace Impurities - Zirconium (Zr) | ≤10.0 ppb |
Particle Count - 0.5 µm and greater | ≤60 par/ml |
Particle Count - 1.0 µm and greater | ≤10 par/ml |
For Microelectronic Use | |
Storage Note: Do not store near Ammonium Hydroxide. White crystals may form around the opening. The crystals can be removed with deionized water. This will not affect the quality of the product in the bottle. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...