Nitric acid 69.0 - 70.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Nitric acid solution

Management of Change (MOC) category = NI

BAKR9601-00 BAKR9601-34 BAKR9601-01 9601-05
CAJT9601-0CS 1016.78 CAD
CAJT9601-0 CAJT9601-34 CAJT9601-1 JT9601-5 CAJT9601-69 CAJT9601-89
Nitric acid 69.0 - 70.0%, BAKER ANALYZED® ACS, J.T.Baker®
Nitric acid

The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.

Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: –42 °C
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (HNO₃) 69.0 - 70.0 %
Arsenic (As) ≤0.010 ppm
Color (APHA) ≤10
Residue after Ignition ≤4 ppm
Chloride (Cl) ≤0.1 ppm
Phosphate (PO₄) ≤0.20 ppm
Sulfate (SO₄) ≤0.5 ppm
Trace Impurities - Aluminum (Al) ≤100.0 ppb
Arsenic and Antimony (as As) ≤4.0 ppb
Trace Impurities - Boron (B) ≤50 ppb
Trace Impurities - Cadmium (Cd), For Information Only
Trace Impurities - Calcium (Ca) ≤200.0 ppb
Trace Impurities - Chromium (Cr) ≤100.0 ppb
Trace Impurities - Copper (Cu) ≤50 ppb
Trace Impurities - Gold (Au) ≤200.0 ppb
Heavy Metals (as Pb) ≤100 ppb
Trace Impurities - Iron (Fe) ≤100.0 ppb
Trace Impurities - Lead (Pb) ≤100 ppb
Trace Impurities - Magnesium (Mg) ≤100.0 ppb
Trace Impurities - Manganese (Mn) ≤100.0 ppb
Trace Impurities - Mercury (Hg) ≤5.0 ppb
Trace Impurities - Nickel (Ni) ≤50 ppb
Trace Impurities - Potassium (K) ≤300.0 ppb
Trace Impurities - Tin (Sn) ≤200.0 ppb
Trace Impurities - Titanium (Ti) ≤200.0 ppb
Trace Impurities - Zinc (Zn) ≤200.0 ppb
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR