Nitric acid 69,0 - 70,0%, BAKER INSTRA-ANALYZED ACS for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Danger
|
Synonyms:
Nitric acid solution
Management of Change (MOC) category = NI
BAKR9598-00
BAKR9598-34
BAKR9598-05
9598-34
BAKR9598-51
CAJT9598-0CS
1358.59
CAD
CAJT9598-0
CAJT9598-34
CAJT9598-5
JT9598-34
CAJT9598-51
Nitric acid 69,0 - 70,0%, BAKER INSTRA-ANALYZED ACS for trace metal analysis, J.T.Baker®
Nitric acid
For trace metal analysis. Lot analysis on label.
Formula:
HNO₃ MW: 63.01 Boiling Pt: 87 Melting Pt: -42 Density: 1.51 Storage Temperature: Ambient |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 EINECS: 231-714-2 UN: 2031 ADR: 8 II Merck Index: 13,06610 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
ACS - Assay (HNO₃) | 69.0 - 70.0 % |
Appearance | Passes Test |
ACS - Color (APHA) | ≤10 |
ACS - Residue after Ignition | ≤2 ppm |
Chloride (Cl) | ≤0.04 ppm |
Phosphate (PO₄) | ≤0.10 ppm |
Sulfate (SO₄) | ≤0.4 ppm |
Trace Impurities - Aluminum (Al) | ≤30.0 ppb |
Arsenic and Antimony (as As) | ≤5.0 ppb |
Trace Impurities - Barium (Ba) | ≤1.0 ppb |
Trace Impurities - Beryllium (Be) | ≤1.0 ppb |
Trace Impurities - Bismuth (Bi) | ≤1.0 ppb |
Trace Impurities - Boron (B) | ≤4.0 ppb |
Trace Impurities - Cadmium (Cd) | ≤1.0 ppb |
Trace Impurities - Calcium (Ca) | ≤50.0 ppb |
Trace Impurities - Chromium (Cr) | ≤10.0 ppb |
Trace Impurities - Cobalt (Co) | ≤1.0 ppb |
Trace Impurities - Copper (Cu) | ≤1.0 ppb |
Trace Impurities - Gallium (Ga) | ≤20.0 ppb |
Trace Impurities - Germanium (Ge) | ≤4.0 ppb |
Trace Impurities - Gold (Au) | ≤4.0 ppb |
Heavy Metals (as Pb) | ≤100 ppb |
Trace Impurities - Iron (Fe) | ≤10.0 ppb |
Trace Impurities - Lead (Pb) | ≤0.5 ppb |
Trace Impurities - Lithium (Li) | ≤1.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤7.0 ppb |
Trace Impurities - Manganese (Mn) | ≤1.0 ppb |
Trace Impurities - Mercury (Hg) | ≤0.5 ppb |
Trace Impurities - Molybdenum (Mo) | ≤5.0 ppb |
Trace Impurities - Nickel (Ni) | ≤1.0 ppb |
Trace Impurities - Niobium (Nb) | ≤1.0 ppb |
Trace Impurities - Potassium (K) | ≤5.0 ppb |
Trace Impurities - Silicon (Si) | ≤20.0 ppb |
Trace Impurities - Silver (Ag) | ≤1.0 ppb |
Trace Impurities - Sodium (Na) | ≤200.0 ppb |
Trace Impurities - Strontium (Sr) | ≤1.0 ppb |
Trace Impurities - Tantalum (Ta) | ≤2.0 ppb |
Trace Impurities - Thallium (Tl) | ≤5.0 ppb |
Trace Impurities - Tin (Sn) | ≤5.0 ppb |
Trace Impurities - Vanadium (V) | ≤1.0 ppb |
Trace Impurities - Zinc (Zn) | ≤5.0 ppb |
Trace Impurities - Zirconium (Zr) | ≤1.0 ppb |
Meets ACS Specifications |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...