Nitric acid 67 - 70%, ULTREX® II Ultrapure Reagent, ultrapure ICP, Trace_metal, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Nitric acid solution

M.O.C. Category N

BAKR6901-05 6901-02 6901-01
CAJT6901-1EA 4971.44 CAD
CAJT6901-1 CAJT6901-2 CAJT6901-5 JT6901-2 JT6901-1
Nitric acid 67 - 70%, ULTREX® II Ultrapure Reagent, ultrapure ICP, Trace_metal, J.T.Baker®
Nitric acid

ULTREX®/ULTREX® II—Acids and salts of extremely high purity with low levels of metallic impurities especially for use in ppt trace element analysis. These products are manufactured using state-of-the-art processing and are packaged under clean room conditions. Post-packaging characterization is performed using advanced analytical instrumental methods. A Certificate of Actual Lot Analysis is provided with every bottle.

Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 I
Merck Index: 13,06610

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (HNO₃)(w/w) 67 - 70 %
Trace Impurities - Aluminum (Al) ≤20 ppt
Trace Impurities - Antimony (Sb) ≤10 ppt
Trace Impurities - Arsenic (As) ≤20 ppt
Trace Impurities - Barium (Ba) ≤10 ppt
Trace Impurities - Beryllium (Be) ≤10 ppt
Trace Impurities - Bismuth (Bi) ≤10 ppt
Trace Impurities - Boron (B) ≤10 ppt
Trace Impurities - Cadmium (Cd) ≤10 ppt
Trace Impurities - Calcium (Ca) ≤10 ppt
Trace Impurities - Cerium (Ce) ≤10 ppt
Trace Impurities - Cesium (Cs) ≤10 ppt
Trace Impurities - Chromium (Cr) ≤10 ppt
Trace Impurities - Cobalt (Co) ≤10 ppt
Trace Impurities - Copper (Cu) ≤10 ppt
Trace Impurities - Dysprosium (Dy) ≤1 ppt
Trace Impurities - Erbium (Er) ≤1 ppt
Trace Impurities - Europium (Eu) ≤1 ppt
Trace Impurities - Gadolinium (Gd) ≤1 ppt
Trace Impurities - Gallium (Ga) ≤10 ppt
Trace Impurities - Germanium (Ge) ≤10 ppt
Trace Impurities - Gold (Au) ≤20 ppt
Trace Impurities - Hafnium (Hf) ≤10 ppt
Trace Impurities - Holmium (Ho) ≤1 ppt
Trace Impurities - Indium (In) ≤1 ppt
Trace Impurities - Iron (Fe) ≤10 ppt
Trace Impurities - Lanthanum (La) ≤1 ppt
Trace Impurities - Lead (Pb) ≤10 ppt
Trace Impurities - Lithium (Li) ≤10 ppt
Trace Impurities - Lutetium (Lu) ≤1 ppt
Trace Impurities - Magnesium (Mg) ≤10 ppt
Trace Impurities - Manganese (Mn) ≤10 ppt
Trace Impurities - Mercury (Hg) ≤50 ppt
Trace Impurities - Molybdenum (Mo) ≤10 ppt
Trace Impurities - Neodymium (Nd) ≤1 ppt
Trace Impurities - Nickel (Ni) ≤20 ppt
Trace Impurities - Niobium (Nb) ≤1 ppt
Trace Impurities - Palladium (Pd) ≤20 ppt
Trace Impurities - Platinum (Pt) ≤20 ppt
Trace Impurities - Potassium (K) ≤10 ppt
Trace Impurities - Praseodymium (Pr) ≤1 ppt
Trace Impurities - Rhenium (Re) ≤10 ppt
Trace Impurities - Rhodium (Rh) ≤10 ppt
Trace Impurities - Rubidium (Rb) ≤10 ppt
Trace Impurities - Ruthenium (Ru) ≤20 ppt
Trace Impurities - Samarium (Sm) ≤1 ppt
Trace Impurities - Scandium (Sc) ≤10 ppt
Trace Impurities - Selenium (Se), For Information Only
Trace Impurities - Silver (Ag) ≤10 ppt
Trace Impurities - Sodium (Na) ≤10 ppt
Trace Impurities - Strontium (Sr) ≤10 ppt
Trace Impurities - Tantalum (Ta), For Information Only
Trace Impurities - Tellurium (Te) ≤1 ppt
Trace Impurities - Terbium (Tb) ≤1 ppt
Trace Impurities - Thallium (Tl) ≤10 ppt
Trace Impurities - Thorium (Th) ≤1 ppt
Trace Impurities - Thulium (Tm) ≤1 ppt
Trace Impurities - Tin (Sn) ≤20 ppt
Trace Impurities - Titanium (Ti) ≤10 ppt
Trace Impurities - Tungsten (W) ≤10 ppt
Trace Impurities - Uranium (U) ≤1 ppt
Trace Impurities - Vanadium (V) ≤10 ppt
Trace Impurities - Ytterbium (Yb) ≤1 ppt
Trace Impurities - Yttrium (Y) ≤1 ppt
Trace Impurities - Zinc (Zn) ≤10 ppt
Trace Impurities - Zirconium (Zr) ≤10 ppt
Certificate Provided Reports Actual Lot Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR