I-CHEM™ Nalgene® Sample Bottles, High-Density Polyethylene, with Caps, Thermo Scientific
Supplier: Thermo Scientific
N301-0125 N301-0250 N301-0500 N311-0125 N311-0250 N311-0500 N311-1000 N319-0125 N319-0500 N319-1000
IRN301-0125CS 684.9 CAD
IRN301-0125 IRN301-0250 IRN301-0500 IRN301-1000 IRN311-0125 IRN311-0250 IRN311-0500 IRN311-1000 IRN319-0125 IRN319-0500 IRN319-1000
I-CHEM™ Nalgene® Sample Bottles, High-Density Polyethylene, with Caps, Thermo Scientific
Bottles Water Sampling Bottles
Narrow mouth and wide mouth bottles, available in natural and amber, are for use in the analyses of metals.
  • Bottles are precleaned
  • Sample-analysis labels are included, unattached
I-CHEM Certified™ 300 series bottles include a Certificate of Analysis that lists detection limits for specified analyses. Manually readable bar-code labels are attached to each bottle with production number for complete traceability. High-density polyethylene bottles have polypropylene caps attached and are guaranteed leakproof for safe sampling, shipping, and storage.
Ordering information: All containers are supplied with caps attached.