VWR® Wide Mouth Round Bottles, HDPE, with Caps
Supplier: VWR International
PLC-03622 PLC-03611
16125-903CS 229.48 CAD
16125-903 36319-343
VWR® Wide Mouth Round Bottles, HDPE, with Caps
Bottles Water Sampling Bottles
Ideal for general laboratory use or for volatile organic analysis.
  • HDPE construction
  • Wide-mouthed design
  • Pre-cleaned in a contaminant-free environment
Precleaned bottles are washed in hot water and laboratory grade non-phosphate detergent, then rinsed three times with tap water and three times with ASTM Type I organic-free water.
They are finally oven dried and cooled to room temperature in an enclosed contaminant-free environment.
Ordering information: Supplied with F-217 polypropylene-lined caps. Pressure-sensitive sample labels are included in each carton.