Nitric acid 68.0 - 70.0%, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ300

NX0409-2 NX0409-4 NX0409P-5 NX0409-75
CANX0409-2EA 194 CAD
CANX0409-2 CANX0409-4 CANX0409P-5 CANX0409-75
Nitric acid 68.0 - 70.0%, GR ACS, Supelco®
Nitric acid

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs.

Formula: HNO₃
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Order Now

Specification Test Results

Aluminum (Al) 0.2ppm max
Appearance To pass test
Arsenic (As) 0.01ppm max
Assay 68.0-70.0%
Calcium (Ca) 0.2ppm max
Chloride (Cl) 0.5ppm max
Chromium (Cr) 0.1ppm max
Color (APHA) 10 max
Copper (Cu) 0.05ppm max
Gold (Au) 0.3ppm max
Heavy metals (as Pb) 0.2ppm max
Iron (Fe) 0.2ppm max
Lead (Pb) 0.1ppm max
Magnesium (Mg) 0.3ppm max
Manganese (Mn) 0.2ppm max
Mercury (Hg) 0.005ppm max
Nickel (Ni) 0.05ppm max
Phosphate (PO4) 0.2ppm max
Potassium (K) 0.5ppm max
Residue after ignition 5ppm max
Sodium (N/A) 0.5ppm max
Sulfate (SO4) 1ppm max
Tin (Sn) 0.3ppm max
Titanium (Ti) 0.3ppm max
Zinc (Zn) 0.3ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR