Nitric acid 67 - 70%, OmniTrace Ultra™ for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

NX0408-1 NX0408-2 NX0408-6 NX0408-7 NX0408-6
CANX0408-1EA 1038 CAD
CANX0408-1 CANX0408-2 CANX0408-6 CANX0408-7 EM-NX0408-6
Nitric acid 67 - 70%, OmniTrace Ultra™ for trace metal analysis, Supelco®

Clear liquid. For low level trace metal analysis. Double-distilled and packaged in ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom conditions. Supplied in specially designed, preleached fluoropolymer resin bottles to guarantee product integrity. Certificate of Analysis supplied with each shipment.

Formula: HNO₃
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,
Merck Index: 13,06608

Order Now

Specification Test Results

Aluminum (Al) 1ppb max
Antimony (Sb) 10pg/g max
Arsenic (As) 20pg/g max
Assay 67-70%
Barium (Ba) 10pg/g max
Beryllium (Be) 10pg/g max
Bismuth (Bi) 10pg/g max
Boron (B) 10pg/g max
Cadmium (Cd) 10pg/g max
Calcium (Ca) 10pg/g max
Cerium (Ce) 10pg/g max
Cesium (Cs) 10pg/g max
Chromium (Cr) 10pg/g max
Cobalt (Co) 10pg/g max
Copper (Cu) 10pg/g max
Dysprosium (Dy) 1pg/g max
Erbium (Er) 1pg/g max
Europium (Eu) 1pg/g max
Gadolinium (Gd) 1pg/g max
Gallium (Ga) 10pg/g max
Germanium (Ge) 10pg/g max
Gold (Au) 20pg/g max
Hafnium (Hf) 10pg/g max
Holmium (Ho) 1pg/g max
Indium (In) 1pg/g max
Iron (Fe) 10pg/g max
Lanthanum (La) 1pg/g max
Lead (Pb) 10pg/g max
Lithium (Li) 10pg/g max
Lutetium (Lu) 1pg/g max
Magnesium (Mg) 10pg/g max
Manganese (Mn) 10pg/g max
Mercury (Hg) 50pg/g max
Molybdenum (Mo) 10pg/g max
Neodymium (Nd) 1pg/g max
Nickel (Ni) 20pg/g max
Niobium (Nb) 1pg/g max
Palladium (Pd) 20pg/g max
Platinum (Pt) 20pg/g max
Potassium (K) 10pg/g max
Praseodymium (Pr) 1pg/g max
Rhenium (Re) 10pg/g max
Rhodium (Rh) 10pg/g max
Rubidium (Rb) 10pg/g max
Ruthenium (Ru) 20pg/g max
Samarium (Sm) 1pg/g max
Scandium (Sc) 10pg/g max
Silver (Ag) 10pg/g max
Sodium (N/A) 10pg/g max
Strontium (Sr) 10pg/g max
Tellurium (Te) 1pg/g max
Terbium (Tb) 1pg/g max
Thallium (Tl) 10pg/g max
Thorium (Th) 1pg/g max
Thulium (Tm) 1pg/g max
Tin (Sn) 20pg/g max
Titanium (Ti) 10pg/g max
Tungsten (W) 10pg/g max
Uranium (U) 1pg/g max
VaN/Adium (V) 10pg/g max
Ytterbium (Yb) 1pg/g max
Yttrium (Y) 1pg/g max
Zinc (Zn) 10pg/g max
Zirconium (Zr) 10pg/g max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR