Nitri-Knit™ Supported Nitrile Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
NK803/8-H5 NK803/11 NK803/7 NK803/9 NK803/10 NK803ES/7 NK803ES/8 NK803ES/9 NK803ES/11 NK803IN/7 NK803IN/8 NK803IN/9 NK803IN/10 NK803IN/11
CA11001-469PK 316.54 CAD
CA11001-469 CA57100-102 CA11001-527 CA57100-100 CA57100-104 CA57100-106 CA89092-984 CA89092-986 CA89092-988 CA89092-990 CA89022-154 CA89022-156 CA89022-158 CA89022-160 CA89022-162
Nitri-Knit™ Supported Nitrile Gloves, Honeywell Safety
Gloves
For use in a wide range of applications, including: automotive assembly and painting; chemical handling and processing; degreasing; dip tank cleanup; fishing industry processing; food processing; printing cleanup; and general refinery, solvent, and maintenance work.
  • High nitrile content offers superior resistance to cuts, snags, and punctures
  • Interlock knit liner adds strength and comfort
  • Treated with permanent antibacterial and anti-mildew agent
  • Color-coded stitching makes size identification fast and easy
  • Compliant with USDA and FDA CFR 21 food processing regulations
  • Length: 30 cm (12")
  • Superior resistance to cuts, snags, and punctures
  • Interlock knit liner adds strength and comfort
  • Treated with permanent antibacterial and anti-mildew agent
  • Color-coded stitching makes size identification fast and easy
  • Compliant with USDA and FDA CFR 21 food-processing regulations
High nitrile content offers superior resistance to cuts, snags, abrasions, and punctures. Gloves provide excellent performance when exposed to solvents, caustics, and animal fats. Gloves feature unique rough texture finish for excellent grip on oily, wet, or dry objects. The interlock knit liner provides added strength and comfort. The nitrile exterior is permanently bonded to the inner liner for added durability. The hand portion of the glove is treated with a permanent antibacterial and anti-mildew agent. The 61 cm (24") extended sleeve gloves provide arm-to-shoulder protection. Gloves contain color-coded stitching for easy identification. Latex-free.
Certifications: Gloves comply with USDA and FDA regulations CFR 21 for use in food processing.