Nickel (II) sulfamate hydrate

Supplier: Thermo Scientific Chemicals
Danger

89020-18 89020-30 89020-A1
CAAA89020-18EA 91.96 CAD
CAAA89020-18 CAAA89020-30 CAAA89020-A1
Nickel (II) sulfamate hydrate
Nickel (II) sulphamate hydrate

Nickel(II) sulfamate is used as an electro plating and surface treating agent. It finds application in metal coloring and casting.


CAS No.: 13770-89-3 Molecular Formula: Ni(SO3NH2)2.xH2O Formula Weight: 250.86(anhy) Physical Form: Crystalline Appearance: Blue Odor: Odorless UN No.: 3077 MDL No.: MFCD00137261

Formula: H₄N₂NiO₆S₂
MW: 250.86 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00137261
CAS Number: 13770-89-3
EINECS: 237-396-1
UN: 3077
ADR: 9 III

Order Now

Specification Test Results

Assay (unspecified) 15.1%-19.2% Ni


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR