Nitric acid 68 - 70%, ARISTAR® ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

BDH3044500MLPC CABDH3130-2.2P BDH3046-2.5L 375000-80659
BDH3044500MLPCCS 1406.36 CAD
BDH3044500MLPC CABDH3130-2.2P BDH3046-2.5L BDH3130-2.5LP
Nitric acid 68 - 70%, ARISTAR® ACS, VWR Chemicals BDH®
Nitric acid

Colorless and free from particulates.

Formula: HNO₃
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Order Now

Specification Test Results

Appearance Colorless, free from susp. matter
Assay (HNO3) 68.0 - 70.0%
Color (APHA) 10 max.
Residue After Ignition 5 ppm max.
Chloride (Cl) 0.5 ppm max.
Sulfate (SO4) 1 ppm max.
Arsenic (As) 0.01 ppm max.
Heavy Metals (as Pb) 0.2 ppm max.
Iron (Fe) 0.2 ppm max.
Manufacture Date Record Actual
Expiration date Record Actual


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR